missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Invitrogen™ ADRBK2 (GRK3) Recombinant Human Protein
ADRBK2, or adrenergic beta receptor kinase 2 is a serine/threonine kinase.
Manufacturer: Invitrogen™ LSPV3974
Description
ADRBK2 is thought to act as a regulator of receptor function.
Specifications
NP_005151.2 | |
Frozen | |
107.2kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Full Length | |
Kinase Assay | |
GRK Kinase Family | |
Yes |
Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis | |
50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol | |
1mg | |
RUO - research use only | |
ADRBK2 (GRK3) | |
Baculovirus | |
Human | |
GST Tag |