Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ ADP-Ribosylation Factor (ARF) 6 Ab-1, Mouse Monoclonal Antibody

Obtain accurate, reproducible results in western blotting experiments with Thermo Scientific™ ADP-Ribosylation Factor (ARF) 6 Ab-1, Mouse Monoclonal Antibody.

$245.65 - $1,405.73

Specifications

Antigen ADP-ribosylation Factor (ARF) 6 Ab-1
Clone 6ARF01
Applications Western Blotting
Classification Monoclonal
Conjugate Unlabeled
View More Specs
Products
Catalog Number Mfr. No. Concentration Quantity Price Quantity    

MS1269P

 
thermo scientific™ lab vision™
MS1269P
200μg/mL 1mL Each for $1,316.92

MS1269P0

 
thermo scientific™ lab vision™
MS1269P0
200μg/mL 100µL Each for $245.65

MS1269PABX

 
thermo scientific™ lab vision™
MS1269PABX
1mg/mL 200µL Each for $1,405.73
Description & Specifications

Specifications

Antigen ADP-ribosylation Factor (ARF) 6 Ab-1
Clone 6ARF01
Applications Western Blotting
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity Human
Format Concentrated
Host Species Mouse
Regulatory Status RUO
Research Discipline G Protein-Coupled Receptors
Type Primary
Immunogen ARF6 protein
Isotype IgG2a kappa

Obtain accurate, reproducible results in western blotting experiments with Thermo Scientific™ ADP-Ribosylation Factor (ARF) 6 Ab-1, Mouse Monoclonal Antibody.

ARF6 (ADP-ribosylation factor 6) is a broadly expressed, differentially regulated GTPase that is present in cytoplasm and on both cell-surface and intracellular membranes and whose functions may include tissue-specific effects on vesicular trafficking during cellular differentiation.

Host Species: Mouse

Clone: 6ARF01

Isotype: IgG1/κ

Species Reactivity: Human. Others not tested.

Epitope: Not determined

Immunogen: ARF6 protein

Ab-1 can be used to examine the subcellular distribution and expression of ARF6 in various tissues and during the differentiation of several well-characterized cell types.

Recommended for:

  • Western Blotting

Note:

USA: RUO; Int'l: RUO