Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ ADP-Ribosylation Factor (ARF) 6 Ab-1, Mouse Monoclonal Antibody


Obtain accurate, reproducible results in western blotting experiments with Thermo Scientific™ ADP-Ribosylation Factor (ARF) 6 Ab-1, Mouse Monoclonal Antibody.

Manufacturer: thermo scientific™ lab vision™  MS1269PABX

 View more versions of this product

Catalog No. MS1269PABX

  • / Each
 

Description & Specifications

Specifications

Host Species Mouse
Immunogen ARF6 protein
Quantity 200µL
Cross Reactivity Human
Antigen ADP-ribosylation Factor (ARF) 6 Ab-1
Format Concentrated
Isotype IgG2a kappa
Applications Western Blotting
Regulatory Status RUO
Clone 6ARF01
Research Discipline G Protein-Coupled Receptors
Type Primary
Classification Monoclonal
Concentration 1mg/mL
Conjugate Unlabeled

Obtain accurate, reproducible results in western blotting experiments with Thermo Scientific™ ADP-Ribosylation Factor (ARF) 6 Ab-1, Mouse Monoclonal Antibody.

ARF6 (ADP-ribosylation factor 6) is a broadly expressed, differentially regulated GTPase that is present in cytoplasm and on both cell-surface and intracellular membranes and whose functions may include tissue-specific effects on vesicular trafficking during cellular differentiation.

Host Species: Mouse

Clone: 6ARF01

Isotype: IgG1/κ

Species Reactivity: Human. Others not tested.

Epitope: Not determined

Immunogen: ARF6 protein

Ab-1 can be used to examine the subcellular distribution and expression of ARF6 in various tissues and during the differentiation of several well-characterized cell types.

Recommended for:

  • Western Blotting

Note:

USA: RUO; Int'l: RUO