Please login to your online account to display your discounted pricing

Adonitol, MP Biomedicals

A crystalline pentose alcohol (C5H12O5) formed by the reduction of ribose which does not reduce benedict solution

$44.66 - $44.66

Chemical Identifiers

Chemical Name or Material Adonitol
CAS 488-81-3
Formula Weight 152.15
MDL Number MFCD00064291
Synonym Adonite
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

ICN10025205

 
mp biomedicals
10025205
5g Each for $44.66
Description & Specifications

Specifications

Formula Weight 152.15
Quantity 5g
Density 1.53g/cm3
EINECS Number 207-685-7
Chemical Name or Material Adonitol
CAS 488-81-3
Synonym Adonite
MDL Number MFCD00064291