Please login to your online account to display your discounted pricing

Alfa Aesar™ Adjuvant Peptide

For cellular and molecular biology applications

Specifications

Product Type Adjuvant Peptide
Molecular Weight 492.48(anhy)
Storage Requirements 2° to 8°C
Sequence N-Acetylmuramyl-L-alanyl-D-isoglutamine
For Use With (Application) Used in immunization to replace more complex proteins. Inhibits the HIV replication in CD4+ H9 lymphocytes
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ62385LB0

 
alfa aesar™
J62385LB0
5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ62385MCR

 
alfa aesar™
J62385MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type Adjuvant Peptide
Molecular Weight 492.48(anhy)
Storage Requirements 2° to 8°C
Sequence N-Acetylmuramyl-L-alanyl-D-isoglutamine
For Use With (Application) Used in immunization to replace more complex proteins. Inhibits the HIV replication in CD4+ H9 lymphocytes
Format Powder
Common Name Muramyl dipeptide
Regulatory Status RUO
Purification Method HPLC