Learn More
Adiponitrile, 99+%, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 043679AE
Description
Adiponitrile is used as an intermediate for the synthesis of hexamethylenediamine (diamino-1,6-hexane). This chemical is an important intermediate for the manufacture of synthetic fiber.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Packaged under Argon
Specifications
Adiponitrile | |
2°C | |
295°C | |
Odorless | |
C6H8N2 | |
MFCD00001975 | |
UN2205 | |
adiponitrile, 1,4-dicyanobutane, adipodinitrile, adipyldinitrile, adipic acid dinitrile, tetramethylene dicyanide, nitrile adipico, tetramethylene cyanide, adipic acid nitrile, adipinsaeuredinitril | |
BTGRAWJCKBQKAO-UHFFFAOYSA-N | |
hexanedinitrile | |
8128 | |
≥99% |
111-69-3 | |
0.951 g/mL | |
163°C (325°F) | |
ChemSeal™ bottles | |
1.438 | |
100 mL | |
1740005 | |
Highly soluble in water (83g/L). | |
C(CCC#N)CC#N | |
108.144 | |
108.14 | |
Liquid |
Chemical Identifiers
111-69-3 | |
108.144 | |
BTGRAWJCKBQKAO-UHFFFAOYSA-N | |
8128 | |
C(CCC#N)CC#N |
C6H8N2 | |
MFCD00001975 | |
adiponitrile, 1,4-dicyanobutane, adipodinitrile, adipyldinitrile, adipic acid dinitrile, tetramethylene dicyanide, nitrile adipico, tetramethylene cyanide, adipic acid nitrile, adipinsaeuredinitril | |
hexanedinitrile |
Safety and Handling
GHS H Statement
H301-H331-H227
Toxic if swallowed.
Toxic if inhaled.
Combustible liquid.
P261-P264b-P270-P271-P301+P310-P304+P340-P312-P330-P501c
H301-H332
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ADIPONITRILE
EINECSNumber : 203-896-3
RTECSNumber : AV2625000
TSCA : Yes
storageNote1 :
- Packaged under Argon
Recommended Storage : Ambient temperatures
RUO – Research Use Only