missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Adipic dihydrazide, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC203315000

 View more versions of this product

Catalog No. AC203315000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Adipic dihydrazide
Plastic bottle
178.0°C to 182.0°C
1071-93-8
100.0
C6H14N4O2
MFCD00007614
adipic acid dihydrazide, adipic acid, dihydrazide, adipic dihydrazide, adipodihydrazide, adipohydrazide, adipyl hydrazide, hexanediohydrazide, hexanedioic acid, 1,6-dihydrazide, hexanedioic acid, dihydrazide, unii-vk98i9yw5m
IBVAQQYNSHJXBV-UHFFFAOYSA-N
hexanedihydrazide
66117
Crystalline Powder
Authentic
White to Yellow
500g
97.5
98%
H2NNHCO(CH2)4CONHNH2
02, I, 277
Solubility in water: soluble. Other solubilities: soluble in acetic acid,slightly soluble in acetone,insoluble in ethanol,ether and benzene
C(CCC(=O)NN)CC(=O)NN
174.2
174.2
98%

Chemical Identifiers

1071-93-8
174.2
IBVAQQYNSHJXBV-UHFFFAOYSA-N
66117
C(CCC(=O)NN)CC(=O)NN
C6H14N4O2
MFCD00007614
adipic acid dihydrazide, adipic acid, dihydrazide, adipic dihydrazide, adipodihydrazide, adipohydrazide, adipyl hydrazide, hexanediohydrazide, hexanedioic acid, 1,6-dihydrazide, hexanedioic acid, dihydrazide, unii-vk98i9yw5m
hexanedihydrazide
Safety and Handling

Safety and Handling

GHS H Statement
May cause an allergic skin reaction.
Toxic to aquatic life with long lasting effects.

GHS P Statement
Wear protective gloves/protective clothing.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation or rash occurs: Get medical advice/attention.

GHS Signal Word: Warning

EINECSNumber : 213-999-5

SDS Documents