Learn More
Adipic dihydrazide, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 203311000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Adipic dihydrazide | |
97.5 | |
178.0°C to 182.0°C | |
Authentic | |
Plastic bottle | |
H2NNHCO(CH2)4CONHNH2 | |
100 g | |
adipic dihydrazide, adipohydrazide, adipic acid dihydrazide, hexanedioic acid, dihydrazide, adipyl hydrazide, hexanediohydrazide, adipic acid, dihydrazide, adipodihydrazide, hexanedioic acid, 1,6-dihydrazide, unii-vk98i9yw5m | |
IBVAQQYNSHJXBV-UHFFFAOYSA-N | |
hexanedihydrazide | |
66117 | |
98% |
1071-93-8 | |
100.0 | |
White to Yellow | |
98% | |
C6H14N4O2 | |
MFCD00007614 | |
02, I, 277 | |
Solubility in water: soluble. Other solubilities: soluble in acetic acid,slightly soluble in acetone,insoluble in ethanol,ether and benzene | |
NNC(=O)CCCCC(=O)NN | |
174.20 | |
174.2 | |
Crystalline Powder |
Chemical Identifiers
1071-93-8 | |
174.20 | |
IBVAQQYNSHJXBV-UHFFFAOYSA-N | |
66117 | |
NNC(=O)CCCCC(=O)NN |
C6H14N4O2 | |
MFCD00007614 | |
adipic dihydrazide, adipohydrazide, adipic acid dihydrazide, hexanedioic acid, dihydrazide, adipyl hydrazide, hexanediohydrazide, adipic acid, dihydrazide, adipodihydrazide, hexanedioic acid, 1,6-dihydrazide, unii-vk98i9yw5m | |
hexanedihydrazide |
Safety and Handling
GHS H Statement
May cause an allergic skin reaction.
Toxic to aquatic life with long lasting effects.
GHS P Statement
Wear protective gloves/protective clothing.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation or rash occurs: Get medical advice/attention.
GHS Signal Word: Warning
EINECSNumber : 213-999-5
RUO – Research Use Only