missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Adipic acid, 99%, Thermo Scientific Chemicals
$52.65 - $116.41
Chemical Identifiers
CAS | 124-04-9 |
---|---|
Molecular Formula | C6H10O4 |
Molecular Weight (g/mol) | 146.142 |
MDL Number | MFCD00004420 |
InChI Key | WNLRTRBMVRJNCN-UHFFFAOYSA-N |
Synonym | adipic acid, adipinic acid, 1,4-butanedicarboxylic acid, adilactetten, acifloctin, acinetten, 1,6-hexanedioic acid, molten adipic acid, kyselina adipova, adipinsaure german |
PubChem CID | 196 |
ChEBI | CHEBI:30832 |
IUPAC Name | hexanedioic acid |
SMILES | C(CCC(=O)O)CC(=O)O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC102815000
|
Thermo Scientific Chemicals
102815000 |
500 g | Plastic bottle |
N/A
|
|
||||
AC102810010
|
Thermo Scientific Chemicals
102810010 |
1 kg | Plastic bottle |
N/A
|
|
||||
AC102810025
|
Thermo Scientific Chemicals
102810025 |
2.5 kg | Plastic bottle |
N/A
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
124-04-9 | |
146.142 | |
WNLRTRBMVRJNCN-UHFFFAOYSA-N | |
196 | |
hexanedioic acid |
C6H10O4 | |
MFCD00004420 | |
adipic acid, adipinic acid, 1,4-butanedicarboxylic acid, adilactetten, acifloctin, acinetten, 1,6-hexanedioic acid, molten adipic acid, kyselina adipova, adipinsaure german | |
CHEBI:30832 | |
C(CCC(=O)O)CC(=O)O |
Specifications
151.0°C to 153.0°C | |
337.0°C | |
100.0 | |
99% | |
HO2C(CH2)4CO2H | |
02, 649 | |
15, 150 | |
WNLRTRBMVRJNCN-UHFFFAOYSA-N | |
hexanedioic acid | |
196 | |
146.14 | |
500 g | |
Authentic | |
Crystalline Powder |
124-04-9 | |
98.5 | |
White | |
C6H10O4 | |
MFCD00004420 | |
01,15 | |
adipic acid, adipinic acid, 1,4-butanedicarboxylic acid, adilactetten, acifloctin, acinetten, 1,6-hexanedioic acid, molten adipic acid, kyselina adipova, adipinsaure german | |
C(CCC(=O)O)CC(=O)O | |
146.142 | |
CHEBI:30832 | |
99% | |
196°C | |
Plastic bottle | |
Adipic acid |
Safety and Handling
GHS H Statement
Causes serious eye damage.
GHS P Statement
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only