missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Adipic acid, 99%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 102810250
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Adipic acid | |
98.5 | |
99% | |
HO2C(CH2)4CO2H | |
02, 649 | |
15, 150 | |
WNLRTRBMVRJNCN-UHFFFAOYSA-N | |
hexanedioic acid | |
196 | |
146.14 | |
196g/mL | |
196°C | |
Fibre drum |
124-04-9 | |
100.0 | |
C6H10O4 | |
MFCD00004420 | |
01,15 | |
adipic acid, adipinic acid, 1,4-butanedicarboxylic acid, adilactetten, acifloctin, acinetten, 1,6-hexanedioic acid, molten adipic acid, kyselina adipova, adipinsaure german | |
C(CCC(=O)O)CC(=O)O | |
146.142 | |
CHEBI:30832 | |
99% | |
25kg | |
Authentic |
Safety and Handling
GHS H Statement
Causes serious eye damage.
GHS P Statement
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO â Research Use Only