missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Adipic acid, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A137050I

 View more versions of this product

Catalog No. AAA137050I


Description

Description

Adipic acid is a monomer used in the production of nylon 66 by polycondenation reaction with hexamethylene diamine. It is also used as a food ingredient and its esters are plasticizers especially in poly vinyl chloride. Further, it reacts with barium hydroxide and undergoes ketonization reaction to get cyclopentanone. In addition to this, it finds application in polyurethane foams, lubricants, adhesives, baking powder, gelling aid and in food flavoring.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Adipic acid is a monomer used in the production of nylon 66 by polycondenation reaction with hexamethylene diamine. It is also used as a food ingredient and its esters are plasticizers especially in poly vinyl chloride. Further, it reacts with barium hydroxide and undergoes ketonization reaction to get cyclopentanone. In addition to this, it finds application in polyurethane foams, lubricants, adhesives, baking powder, gelling aid and in food flavoring.

Solubility
Soluble in methanol and acetone. Slightly soluble in cyclohexane. Insoluble in benzene, acetic acid and petroleum ether.

Notes
Incompatible with oxidizing agents
Specifications

Specifications

Adipic acid
151°C to 155°C
∼205°C (3mmHg)
MFCD00004420
14,162
Soluble in methanol and acetone. Slightly soluble in cyclohexane. Insoluble in benzene,acetic acid and petroleum ether.
C(CCC(=O)O)CC(=O)O
146.142
CHEBI:30832
99%
5000 g
Odorless
2.7
124-04-9
C6H10O4
1209788
adipic acid, adipinic acid, 1,4-butanedicarboxylic acid, adilactetten, acifloctin, acinetten, 1,6-hexanedioic acid, molten adipic acid, kyselina adipova, adipinsaure german
WNLRTRBMVRJNCN-UHFFFAOYSA-N
hexanedioic acid
196
146.14
1.36
196°C (384°F)

Chemical Identifiers

124-04-9
146.142
WNLRTRBMVRJNCN-UHFFFAOYSA-N
196
hexanedioic acid
C6H10O4
MFCD00004420
adipic acid, adipinic acid, 1,4-butanedicarboxylic acid, adilactetten, acifloctin, acinetten, 1,6-hexanedioic acid, molten adipic acid, kyselina adipova, adipinsaure german
CHEBI:30832
C(CCC(=O)O)CC(=O)O
Safety and Handling

Safety and Handling

P280-P305+P351+P338-P310

H318

EINECSNumber : 204-673-3

RTECSNumber : AU8400000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only