Please login to your online account to display your discounted pricing

Alfa Aesar™ Adipamide, 98%

$40.01 - $40.01

Chemical Identifiers

Chemical Name or Material Adipamide
CAS 628-94-4
Molecular Formula C6H12N2O2
Linear Formula H2NCO(CH2)4CONH2
Formula Weight 144.17g/mol
MDL Number MFCD00008045
Synonym Butane-1,4-dicarboxamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2425206

 
alfa aesar™
B24252-06
5g Each for $40.01
Description & Specifications

Specifications

Melting Point 224° to 228°C
Quantity 5g
Chemical Name or Material Adipamide
CAS 628-94-4
Assay 98%
Molecular Formula C6H12N2O2
Linear Formula H2NCO(CH2)4CONH2
Formula Weight 144.17g/mol
MDL Number MFCD00008045
Synonym Butane-1,4-dicarboxamide
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
TSCA TSCA
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.