Please login to your online account to display your discounted pricing
Thermo Scientific™ Adhesive Plate Seals
Ensure sample integrity with these Adhesive Plate Seals, an effective sealing method.
Manufacturer: thermo scientific™  AB1450
 View more versions of this product
Catalog No. AB-1450
  • / Pack of 50 Each
Description & Specifications
Specifications
Adhesive Adhesive Plate Seal
Applications Long- or short-term sample storage
Temperature Range -70°C to 110°C
No. per Case 50
Description Can apply over pierced foil seals for storage
No. per Pack 50
Adhesive Plate Seals are more convenient and cost effective than other sealing methods, and are available in a wide range of styles to suit any application. Thermo Scientific adhesive seals do not leave behind a sticky residue when the seal is removed, allowing the plate to be sealed multiple times.