Thermo Scientific™ Adhesive PCR Plate Seals
Prevent sample evaporation during PCR, thermal cycling, or sample storage with this adhesive film.
$270.92 - $401.31
Specifications
For Use With PCR and qPCR
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Description Price Quantity  
Catalog Number Mfr. No. Description Price Quantity  
AB-0558 Thermo Scientific™
AB0558
Peelable adhesive PCR film Pack of 100 Each for $398.50
AB-0626 Thermo Scientific™
AB0626
Peelable adhesive PCR foil Pack of 100 Each for $401.31
AB-1170 Thermo Scientific™
AB1170
ABsolute QPCR Seal Pack of 50 Each for $270.92
Specifications
Description
Adhesive PCR Plate Seals are a reliable method of sealing plates for PCR applications. The high integrity sealing materials give the best protection against evaporation for high efficiency PCR.
Highlights
  • Clear for sample inspection
  • Strong adhesive prevents evaporation during thermal cycling or sample storage
  • No sticky residue on the plate when the seal is peeled off
  • End tabs for easy seal application and removal
  • Recommended for use in conjunction with a cycler that has a screw-down or clip-down lid
  • Temperature range: -20°C to 120°C
Description & Specifications
Specifications
PCR and qPCR