Please login to your online account to display your discounted pricing

Thermo Scientific™ Adhesive PCR Plate Seals - PROMO

Prevent sample evaporation during PCR, thermal cycling, or sample storage with this adhesive film.

$160.76 - $235.73

Adhesive Strong
View More Specs
Products
Catalog Number Mfr. No. Applications Product Type Temperature Range Price Quantity    

AB0558PM

 
thermo scientific™
AB0558PM
PCR Thermal cycling with a screw-down or clip-down lid; sample storage Peelable adhesive PCR Film -20 to +120°C Each for $235.73

AB0626PM

 
thermo scientific™
AB0626PM
PCR Thermal cycling with a screw-down or clip-down lid; sample storage Peelable adhesive PCR Foil -40°to +120°C Pack of 100 Each for $235.73

AB1170PM

 
thermo scientific™
AB1170PM
For qPCR and other fluorescence applications ABsolute QPCR Seal –80°C to 110°C Each for $160.76
Description & Specifications

Specifications

Adhesive Strong
Plate Compatibility 96-Well Plates, 96-Well Plate, Fast (0.1 ml), 384-Well Plates
Color Clear
Pierceable Yes
For Use With PCR and qPCR

  • Clear for sample inspection
  • Strong adhesive prevents evaporation during thermal cycling or sample storage
  • No sticky residue on the plate when the seal is peeled off
  • End tabs for easy seal application and removal
  • Recommended for use in conjunction with a cycler that has a screw-down or clip-down lid
  • Temperature range: -20°C to 120°C