Please login to your online account to display your discounted pricing

Thermo Scientific™ Adhesive PCR Plate Seals - PROMO


Prevent sample evaporation during PCR, thermal cycling, or sample storage with this adhesive film.

Manufacturer: thermo scientific™  AB1170PM

 View more versions of this product

Catalog No. AB1170PM

  • / Each
 

Description & Specifications

Specifications

Product Type ABsolute QPCR Seal
Adhesive Strong
Plate Compatibility 96-Well Plates, 96-Well Plate, Fast (0.1 ml), 384-Well Plates
Applications For qPCR and other fluorescence applications
Color Clear
Pierceable Yes
Temperature Range –80°C to 110°C
For Use With PCR and qPCR