Please login to your online account to display your discounted pricing

Thermo Scientific™ Adhesive PCR Plate Seals - PROMO


Prevent sample evaporation during PCR, thermal cycling, or sample storage with this adhesive film.

Manufacturer: thermo scientific™  AB0626PM

 View more versions of this product

Catalog No. AB0626PM

  • / Pack of 100 Each
 

Description & Specifications

Specifications

Product Type Peelable adhesive PCR Foil
Plate Compatibility 96-Well Plates, 96-Well Plate, Fast (0.1 ml), 384-Well Plates
Applications PCR Thermal cycling with a screw-down or clip-down lid; sample storage
Color Clear
Pierceable Yes
For Use With PCR and qPCR
Adhesive Strong
Temperature Range -40°to +120°C