Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Adenovirus Type 5 E1A Ab-1, Mouse Monoclonal Antibody

Obtain accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence, and immunoprecipitation experiments.

$239.59 - $1,256.76

Specifications

Antigen Adenovirus Type 5 E1A Ab-1
Clone M58
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blotting
View More Specs
Products
Catalog Number Mfr. No. Concentration Conjugate Quantity Price Quantity    

MS587B

 
thermo scientific™ lab vision™
MS587B
200μg/mL Biotin 1mL Each for $1,256.76

MS587B0

 
thermo scientific™ lab vision™
MS587B0
200μg/mL Biotin 100µL Each for $333.10

MS587B1

 
thermo scientific™ lab vision™
MS587B1
200μg/mL Biotin 500µL Each for $875.27

MS587P

 
thermo scientific™ lab vision™
MS587P
200μg/mL Unlabeled 1mL Each for $964.59

MS587P0

 
thermo scientific™ lab vision™
MS587P0
200μg/mL Unlabeled 100µL Each for $239.59

MS587P1

 
thermo scientific
MS587P1
200μg/mL Unlabeled 500µL Each for $672.20

MS587P1ABX

 
thermo scientific™ lab vision™
MS587P1ABX
1mg/mL Unlabeled 100µL Each for $746.43

MS587PABX

 
thermo scientific™ lab vision™
MS587PABX
1mg/mL Unlabeled 200µL Each for $1,047.24
Description & Specifications

Specifications

Antigen Adenovirus Type 5 E1A Ab-1
Clone M58
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blotting
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blotting
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blotting
Classification Monoclonal
Cross Reactivity Adenovirus
Format Concentrated
Host Species Mouse
Regulatory Status RUO
Research Discipline Infectious Disease
Type Primary
Immunogen Adenovirus
Isotype IgG2a kappa

Obtain accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence, and immunoprecipitation experiments with Thermo Scientific™ Adenovirus Type 5 E1A Ab-1, Mouse Monoclonal Antibody.

 

The early region (E1A) of the adenovirus genome plays a central role in cellular transformation and regulation of gene expression. The E1A region encodes a series of related proteins (35-46kDa) with multifuctional capabilities that form a specific complex with the retinoblastoma tumor suppressor gene product (Rb protein). The E1A and E1B regions together comprise the transforming region of adenovirus. While expression of E1A alone is sufficient to immortalize primary cells, complete transformation requires the additional expression of the E1B region. Several conserved regions of E1A are similar to portions of other viral oncoproteins like the HPV-16 and HPV-18 E7 and SV40 large T antigen.

Host Species: Mouse

Clone: M58

Isotype: IgG2a/κ

Species Reactivity: Adenovirus.

Immunogen: Adenovirus

Molecular Weight: 35-46kDa

Positive Control: 293 cells or adenovirus infected cells and tissues

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native verified)

Note:

USA: RUO; Int'l: RUO