missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Adenovirus Type 5 E1A Ab-1, Mouse Monoclonal Antibody

Obtain accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence, and immunoprecipitation experiments.

Manufacturer:  Thermo Scientific™ Lab Vision™ MS587P1ABX

 View more versions of this product

Catalog No. MS587P1ABX


 

Specifications

Description

Obtain accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence, and immunoprecipitation experiments with Thermo Scientific™ Adenovirus Type 5 E1A Ab-1, Mouse Monoclonal Antibody.

 

The early region (E1A) of the adenovirus genome plays a central role in cellular transformation and regulation of gene expression. The E1A region encodes a series of related proteins (35-46kDa) with multifuctional capabilities that form a specific complex with the retinoblastoma tumor suppressor gene product (Rb protein). The E1A and E1B regions together comprise the transforming region of adenovirus. While expression of E1A alone is sufficient to immortalize primary cells, complete transformation requires the additional expression of the E1B region. Several conserved regions of E1A are similar to portions of other viral oncoproteins like the HPV-16 and HPV-18 E7 and SV40 large T antigen.

Host Species: Mouse

Clone: M58

Isotype: IgG2a/κ

Species Reactivity: Adenovirus.

Immunogen: Adenovirus

Molecular Weight: 35-46kDa

Positive Control: 293 cells or adenovirus infected cells and tissues

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native verified)

Note:

USA: RUO; Int'l: RUO

Description & Specifications

Specifications

Adenovirus Type 5 E1A Ab-1
Monoclonal
1mg/mL
Concentrated
Adenovirus
100μL
Infectious Disease
Monoclonal
Immunofluorescence,Immunoprecipitation,Western Blotting
M58
Unlabeled
Murine
IgG2a kappa
RUO
Primary
Adenovirus