missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Adenovirus Type 2 E1A Ab-2, Mouse Monoclonal Antibody

Obtain accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence, and immunoprecipitation experiments.

$243.18 - $1,062.00

Specifications

Antigen Adenovirus Type 2 E1A Ab-2
Clone M73
Applications Immunofluorescence,Immunoprecipitation,Western Blotting,Immunofluorescence,Immunoprecipitation,Western Blotting,Immunofluorescence,Immunoprecipitation,Western Blotting
Classification Monoclonal
Conjugate Unlabeled
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Concentration Quantity Price Quantity  
Catalog Number Mfr. No. Concentration Quantity Price Quantity  
MS588P Thermo Scientific™ Lab Vision™
MS588P
200μg/mL 1mL Each for $978.18
MS588P0 Thermo Scientific™ Lab Vision™
MS588P0
200μg/mL 100μL Each for $243.18
MS588P1 Thermo Scientific™ Lab Vision™
MS588P1
200μg/mL 500μL Each for $681.65
MS588P1ABX Thermo Scientific™ Lab Vision™
MS588P1ABX
1mg/mL 100μL Each for $756.94
MS588PABX Thermo Scientific™ Lab Vision™
MS588PABX
1mg/mL 200μL Each for $1,062.00
Specifications

Description

Obtain accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence, and immunoprecipitation experiments with Thermo Scientific™ Adenovirus Type 2 E1A Ab-2, Mouse Monoclonal Antibody.

The adenovirus early gene product E1A is a potent stimulator of cellular proliferation, which when overexpressed can overcome the growth-inhibitory effects of TGF-β. The E1A region encodes a series of related proteins (35-46kDa) with multifuctional capabilities and form a specific complex with the retinoblastoma tumor suppressor gene product (Rb protein). The E1A and E1B regions together comprise the transforming region of adenovirus. While expression of E1A alone is sufficient to immortalize primary cells, complete transformation requires the additional expression of the E1B region.

Host Species: Mouse

Clone: M73

Isotype: IgG2a/κ

Species Reactivity: Adenovirus.

Immunogen: Adenovirus

Molecular Weight: 35-46kDa

Positive Control: 293 cells or adenovirus infected tissues

Cellular Localization: Nuclear.

Ab-2 immunoprecipitates E1A associated proteins.

Recommended for:

  • Immunohistochemistry
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native verified)

Note:

USA: RUO; Int'l: RUO

Description & Specifications

Specifications

Adenovirus Type 2 E1A Ab-2
Immunofluorescence,Immunoprecipitation,Western Blotting,Immunofluorescence,Immunoprecipitation,Western Blotting,Immunofluorescence,Immunoprecipitation,Western Blotting
Unlabeled
Murine
Infectious Disease
Adenovirus
M73
Monoclonal
Concentrated
RUO
Primary
IgG2a kappa