Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Adenovirus Type 2 E1A Ab-2, Mouse Monoclonal Antibody


Obtain accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence, and immunoprecipitation experiments.

Manufacturer: thermo scientific™ lab vision™  MS588P1ABX

 View more versions of this product

Catalog No. MS588P1ABX

  • / Each
 

Description & Specifications

Specifications

Format Concentrated
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blotting
Immunogen Adenovirus
Isotype IgG2a kappa
Clone M73
Quantity 100µL
Regulatory Status RUO
Cross Reactivity Adenovirus
Antigen Adenovirus Type 2 E1A Ab-2
Host Species Mouse
Classification Monoclonal
Conjugate Unlabeled
Research Discipline Infectious Disease
Type Primary
Concentration 1mg/mL

Obtain accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence, and immunoprecipitation experiments with Thermo Scientific™ Adenovirus Type 2 E1A Ab-2, Mouse Monoclonal Antibody.

The adenovirus early gene product E1A is a potent stimulator of cellular proliferation, which when overexpressed can overcome the growth-inhibitory effects of TGF-β. The E1A region encodes a series of related proteins (35-46kDa) with multifuctional capabilities and form a specific complex with the retinoblastoma tumor suppressor gene product (Rb protein). The E1A and E1B regions together comprise the transforming region of adenovirus. While expression of E1A alone is sufficient to immortalize primary cells, complete transformation requires the additional expression of the E1B region.

Host Species: Mouse

Clone: M73

Isotype: IgG2a/κ

Species Reactivity: Adenovirus.

Immunogen: Adenovirus

Molecular Weight: 35-46kDa

Positive Control: 293 cells or adenovirus infected tissues

Cellular Localization: Nuclear Ab-2 immunoprecipitates E1A associated proteins.

Recommended for:

  • Immunohistochemistry
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native verified)

Note:

USA: RUO; Int'l: RUO