Please login to your online account to display your discounted pricing

Adenosine, Free Base, Cell Culture Reagent, 99.8%, For HPLC analysis, MP Biomedicals™


Quantity 25g

Manufacturer: mp biomedicals  19460925

 View more versions of this product

Catalog No. ICN19460925

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Powder
Assay 99.80%
Formula Weight 267.2
Color White
Quantity 25g
Chemical Name or Material Adenosine
Name Note For HPLC analysis
Molecular Formula C10H13N5O4
CAS 58-61-7
Synonym 9-b-D-Ribofuranosyladenine

Specifications

Chemical Name or Material Adenosine
Name Note For HPLC analysis
CAS 58-61-7
Physical Form Powder
Molecular Formula C10H13N5O4
Formula Weight 267.2
Synonym 9-b-D-Ribofuranosyladenine