Please login to your online account to display your discounted pricing

Adenosine, Free Base, Cell Culture Reagent, 99.8%, For HPLC analysis, MP Biomedicals™


Quantity 1g

Manufacturer: mp biomedicals  0219460901

 View more versions of this product

Catalog No. ICN19460901

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 1g
Chemical Name or Material Adenosine
Name Note For HPLC analysis
CAS 58-61-7
Purity Grade 99.80%
Physical Form White powder
Molecular Formula C10H13N5O4
Formula Weight 267.2g/mol
Synonym 9-b-D-Ribofuranosyladenine

Specifications

Chemical Name or Material Adenosine
Name Note For HPLC analysis
CAS 58-61-7
Purity Grade 99.80%
Physical Form White powder
Molecular Formula C10H13N5O4
Formula Weight 267.2g/mol
Synonym 9-b-D-Ribofuranosyladenine