Please login to your online account to display your discounted pricing

Adenosine, Free Base, Cell Culture Reagent, 99.8%, For HPLC analysis, MP Biomedicals™

$145.68 - $145.68

Chemical Identifiers

Chemical Name or Material Adenosine
Name Note For HPLC analysis
CAS 58-61-7
Physical Form Powder
Molecular Formula C10H13N5O4
Formula Weight 267.2
Synonym 9-b-D-Ribofuranosyladenine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

ICN19460925

 
mp biomedicals
19460925
25g Each for $145.68
Description & Specifications

Specifications

Physical Form Powder
Assay 99.80%
Formula Weight 267.2
Color White
Quantity 25g
Chemical Name or Material Adenosine
Name Note For HPLC analysis
Molecular Formula C10H13N5O4
CAS 58-61-7
Synonym 9-b-D-Ribofuranosyladenine