Please login to your online account to display your discounted pricing

Adenosine 5'-triphosphate, disodium salt hydrate, 98%, ACROS Organics™


Quantity 10g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC102800100

 View more versions of this product

Catalog No. AC102800100

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥97.5% (HPLC)
Physical Form Liquid
Formula Weight 551.14
Quantity 10g
Color Clear colorless to yellow to slightly brown
Water 10% Max. (K.F.)
pH 2.5 to 3.5 (5% aq. soln.)
CAS Max % 100.0
Heavy Metals (as Pb) 20ppm max.
Ratio of Absorbances 0.76 to 0.8 (A250/A260) 0.13 to 0.17 (A280/A260)
Chemical Name or Material Adenosine 5'-triphosphate, disodium salt hydrate
Infrared Spectrum Authentic
Packaging Glass bottle
Related Substances 2.0% max. AMP + ADP (HPLC)
Assay Percent Range 98%
Molecular Formula C10H14N5Na2O13P3·xH2O
Spectrophotometry 98 to 102% (lambda max. 259nm, pH 7) molar absorptivity 15400 ±2%, on dried substance
Appearance of Solution (5% aq. soln.) clear, colorless
CAS 34369-07-8
Synonym ATP, disodium salt hydrate
Bacterial Endotoxins (LAL−test) negative reaction (< 0.6 EU/mg)
CAS Min % 97.5

Specifications

Chemical Name or Material Adenosine 5'-triphosphate, disodium salt hydrate
CAS 34369-07-8
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5% (HPLC)
Assay Percent Range 98%
Molecular Formula C10H14N5Na2O13P3·xH2O
Formula Weight 551.14
Synonym ATP, disodium salt hydrate