missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Adenosine 5'-monophosphate sodium salt hydrate, 99%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC440710050
Specifications
Adenosine 5′-monophosphate sodium salt hydrate | |
98.5 | |
98.5% min. (HPLC) | |
5'-adenylic acid,sodium salt, hydrate 9ci, adenosine 5'-monophosphate sodium salt, adenosine 5-monophosphoric acid disodium salt, disodium hydrate adenosine-5-monophosphate 2-, pubchem14181, sodium 2r,3s,4r,5r-5-6-amino-9h-purin-9-yl-3,4-dihydroxytetrahydrofuran-2-yl methyl phosphate hydrate, sodium 2r,3s,4r,5r-5-6-amino-9h-purin-9-yl-3,4-dihydroxytetrahydrofuran-2-yl methyl phosphate hydrate 2:1:x, sodium 2r,3s,4r,5r-5-6-amino-9h-purin-9-yl-3,4-dihydroxytetrahydrofuran-2-yl methyl phosphate hydrate x:1:x | |
C1=NC2=C(C(=N1)N)N=CN2C3C(C(C(O3)COP(=O)(O)[O-])O)O.[Na+] | |
369.21 | |
369.21 | |
99% | |
Glass bottle | |
White | |
5g |
149022-20-8 | |
100.0 | |
C10H13N5NaO7P·xH2O | |
FYWLYWMEGHCZAX-MCDZGGTQSA-M | |
sodium;[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl hydrogen phosphate | |
23674992 | |
Powder | |
Authentic | |
20% Max. | |
4.50 to 6.50 |
Chemical Identifiers
149022-20-8 | |
369.21 | |
5'-adenylic acid,sodium salt, hydrate 9ci, adenosine 5'-monophosphate sodium salt, adenosine 5-monophosphoric acid disodium salt, disodium hydrate adenosine-5-monophosphate 2-, pubchem14181, sodium 2r,3s,4r,5r-5-6-amino-9h-purin-9-yl-3,4-dihydroxytetrahydrofuran-2-yl methyl phosphate hydrate, sodium 2r,3s,4r,5r-5-6-amino-9h-purin-9-yl-3,4-dihydroxytetrahydrofuran-2-yl methyl phosphate hydrate 2:1:x, sodium 2r,3s,4r,5r-5-6-amino-9h-purin-9-yl-3,4-dihydroxytetrahydrofuran-2-yl methyl phosphate hydrate x:1:x | |
sodium;[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl hydrogen phosphate |
C10H13N5NaO7P·xH2O | |
FYWLYWMEGHCZAX-MCDZGGTQSA-M | |
23674992 | |
C1=NC2=C(C(=N1)N)N=CN2C3C(C(C(O3)COP(=O)(O)[O-])O)O.[Na+] |