missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Adenosine 5'-monophosphate, 99%, ACROS Organics™

Manufacturer:  Acros Organics 102790250

 View more versions of this product

Catalog No. AC102790250



Specifications

Specifications

0.76 to 0.80 (A250/A260), 0.14 to 0.16 (A280/A260)
Authentic
Glass bottle
95.0% min. (c=1, 0.2N NaOH, 430nm) 1cm cell
178.0°C to 185.0°C
61-19-8
100.0
C10H14N5O7P
26, IV, 3615
adenosine 5'-monophosphate, 5'-adenylic acid, adenosine monophosphate, adenosine phosphate, adenylic acid, adenylate, phosphaden, 5'-amp, adenosine 5'-phosphate, phosphentaside
UDMBCSSLTHHNCD-KQYNXXCUSA-N
[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl dihydrogen phosphate
6083
347.21
99%
Adenosine 5′-monophosphate
6% max. (105°C, 5 hrs)
98.0% min. (lambda min. 260nm, pH 7) molar absorptivity 15400 +/- 2% min., on dry basis
Colorless to White
25g
98.5
99%
MFCD00005750
15, 146
Solubility in water: soluble. Other solubilities: readily soluble in boiling water, soluble in ethanol
C1=NC2=C(C(=N1)N)N=CN2C3C(C(C(O3)COP(=O)(O)O)O)O
347.224
CHEBI:16027
Crystalline Powder

Chemical Identifiers

61-19-8
347.224
UDMBCSSLTHHNCD-KQYNXXCUSA-N
6083
[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl dihydrogen phosphate
C10H14N5O7P
MFCD00005750
adenosine 5'-monophosphate, 5'-adenylic acid, adenosine monophosphate, adenosine phosphate, adenylic acid, adenylate, phosphaden, 5'-amp, adenosine 5'-phosphate, phosphentaside
CHEBI:16027
C1=NC2=C(C(=N1)N)N=CN2C3C(C(C(O3)COP(=O)(O)O)O)O
SDS Documents