Please login to your online account to display your discounted pricing

Adenosine-5'-diphosphate monosodium salt, >95%, MP Biomedicals™


Quantity 5g

Manufacturer: mp biomedicals  15026005

 View more versions of this product

Catalog No. ICN15026005

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Powder
Formula Weight 449.2 (Anhydrous)
Color White
Quantity 5g
Assay Percent Range >95%
Chemical Name or Material Adenosine-5'-diphosphate monosodium salt
Molecular Formula C10H14N5O10P2Na·+H2O
CAS 20398-34-9
Synonym ADP-Na

Specifications

Chemical Name or Material Adenosine-5'-diphosphate monosodium salt
CAS 20398-34-9
Physical Form Powder
Assay Percent Range >95%
Molecular Formula C10H14N5O10P2Na·+H2O
Formula Weight 449.2 (Anhydrous)
Synonym ADP-Na