Please login to your online account to display your discounted pricing

Adenosine-5'-diphosphate monosodium salt, >95%, MP Biomedicals™


Quantity 1g

Manufacturer: mp biomedicals  15026001

 View more versions of this product

Catalog No. ICN15026001

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 449.2 (Anhydrous)
Physical Form Powder
Color White
Quantity 1g
CAS 20398-34-9
Chemical Name or Material Adenosine-5'-diphosphate monosodium salt
Assay Percent Range >95%
Molecular Formula C10H14N5O10P2Na·+H2O
Synonym ADP-Na

Specifications

Chemical Name or Material Adenosine-5'-diphosphate monosodium salt
CAS 20398-34-9
Physical Form Powder
Assay Percent Range >95%
Molecular Formula C10H14N5O10P2Na·+H2O
Formula Weight 449.2 (Anhydrous)
Synonym ADP-Na