Please login to your online account to display your discounted pricing

Adenosine-5'-diphosphate monosodium salt, >95%, MP Biomedicals™

$111.70 - $444.84

Chemical Identifiers

Chemical Name or Material Adenosine-5'-diphosphate monosodium salt
CAS 20398-34-9
Physical Form Powder
Assay Percent Range >95%
Molecular Formula C10H14N5O10P2Na·+H2O
Formula Weight 449.2 (Anhydrous)
Synonym ADP-Na
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

ICN15026005

 
mp biomedicals
15026005
5g Each for $444.84

ICN15026001

 
mp biomedicals
15026001
1g Each for $111.70
Description & Specifications

Specifications

Physical Form Powder
Formula Weight 449.2 (Anhydrous)
Color White
Quantity 5g
Assay Percent Range >95%
Chemical Name or Material Adenosine-5'-diphosphate monosodium salt
CAS 20398-34-9
Molecular Formula C10H14N5O10P2Na·+H2O
Synonym ADP-Na