missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Adenosine 2',3'-cyclic monophosphate, sodium salt hydrate, 97%, Acros Organics

Manufacturer:  Thermo Fisher Scientific 228212500

 View more versions of this product

Catalog No. AC228212500


 

Specifications Description & Specifications

Specifications

Adenosine 2', 3'-cyclic monophosphate, sodium salt hydrate
37063-35-7
100.0
258 to 262nm
13000L/(mole.cm) (lambda min., pH 7) on dry substance
97%
0.76 to 0.82 (A250/A260), 0.13 to 0.17 (A280/A260)
MFCD00149366
White
97%
96.0
Authentic
Powder
≥96.0%
C10H11N5NaO6P·xH2O
351.19
8% max. (K.F.)
250mg

Chemical Identifiers

37063-35-7
100.0
C10H11N5NaO6P·xH2O
MFCD00149366
≥96.0%
96.0
97%
351.19
Adenosine 2', 3'-cyclic monophosphate, sodium salt hydrate, 97%

Safety and Handling

SDS & Safety Documents

EINECSNumber : 253-328-3