Please login to your online account to display your discounted pricing

Adenosine 2',3'-cyclic monophosphate, sodium salt hydrate, 97%, Acros Organics

$481.55 - $481.55

Chemical Identifiers

Chemical Name or Material Adenosine 2', 3'-cyclic monophosphate, sodium salt hydrate
Name Note 97%
CAS 37063-35-7
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C10H11N5NaO6P·xH2O
Formula Weight 351.19
MDL Number MFCD00149366
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC228212500

  SDS
thermo fisher scientific
228212500
250mg Each for $481.55
Description & Specifications

Specifications

Assay ≥96.0%
Formula Weight 351.19
Physical Form Powder
Quantity 250mg
Color White
EINECS Number 253-328-3
CAS Max % 100.0
CAS Min % 96.0
CAS 37063-35-7
Lambda Max 258 to 262nm
Molar Absorption Coefficient 13000L/(mole.cm) (lambda min., pH 7) on dry substance
Name Note 97%
Molecular Formula C10H11N5NaO6P·xH2O
MDL Number MFCD00149366
Assay Percent Range 97%
Chemical Name or Material Adenosine 2', 3'-cyclic monophosphate, sodium salt hydrate
Ratio of Absorbances 0.76 to 0.82 (A250/A260), 0.13 to 0.17 (A280/A260)
Water 8% max. (K.F.)
Infrared Spectrum Authentic