Please login to your online account to display your discounted pricing

Adenine Sulfate Dihydrate, Hemisulfate, 99.66%, For HPLC analysis, MP Biomedicals™


Quantity 5g

Manufacturer: mp biomedicals  19460705

 View more versions of this product

Catalog No. ICN19460705

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Powder
Formula Weight 404.36
Color White to off-white
Quantity 5g
Chemical Name or Material Adenine Sulfate Dihydrate, Hemisulfate
Name Note For HPLC analysis
Assay Percent Range 0.9966
Molecular Formula (C5H5N5)2·H2SO4·2H2O
CAS 6509-19-9

Specifications

Chemical Name or Material Adenine Sulfate Dihydrate, Hemisulfate
Name Note For HPLC analysis
CAS 6509-19-9
Physical Form Powder
Assay Percent Range 0.9966
Molecular Formula (C5H5N5)2·H2SO4·2H2O
Formula Weight 404.36