Please login to your online account to display your discounted pricing

Adenine Sulfate Dihydrate, Hemisulfate, 99.66%, For HPLC analysis, MP Biomedicals™

$66.19 - $189.82

Chemical Identifiers

Chemical Name or Material Adenine Sulfate Dihydrate, Hemisulfate
Name Note For HPLC analysis
CAS 6509-19-9
Physical Form Powder
Assay Percent Range 0.9966
Molecular Formula (C5H5N5)2·H2SO4·2H2O
Formula Weight 404.36
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

ICN19460705

 
mp biomedicals
19460705
5g Each for $66.19

ICN19460725

 
mp biomedicals
19460725
25g Each for $189.82
Description & Specifications

Specifications

Physical Form Powder
Formula Weight 404.36
Color White to off-white
Quantity 5g
Chemical Name or Material Adenine Sulfate Dihydrate, Hemisulfate
Name Note For HPLC analysis
Assay Percent Range 0.9966
Molecular Formula (C5H5N5)2·H2SO4·2H2O
CAS 6509-19-9