Please login to your online account to display your discounted pricing

Adenine hydrochloride, 99.20%, MP Biomedicals™


A purine nucleobase widespread throughout animal and plant tissues combined with niacinamide, D-ribose, and phosphoric acids; a constituent of nucleic acids and coenzymes, such as codehydrase I and II, adenylic acid, coalaninedehydrase

Quantity 25g

Manufacturer: mp biomedicals  0210019125

 View more versions of this product

Catalog No. ICN10019125

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 25g
Chemical Name or Material Adenine hydrochloride
CAS 6055-72-7
Physical Form White to off-white powder
Assay Percent Range 0.992
Molecular Formula C5H5N5· HCl
Formula Weight 171.6g/mol
MDL Number MFCD00150864
Synonym 6-Amino-1H-purine

Specifications

Chemical Name or Material Adenine hydrochloride
CAS 6055-72-7
Physical Form White to off-white powder
Assay Percent Range 0.992
Molecular Formula C5H5N5· HCl
Formula Weight 171.6g/mol
MDL Number MFCD00150864
Synonym 6-Amino-1H-purine