Please login to your online account to display your discounted pricing
Thermo Scientific™ Adapters for Single Blood Bag Bucket with Filter Pack for Thermo Scientific™ HAEMAFlex™ 6/8 Rotors
Accommodate 450–550mL Triple, Quad or Quint blood bags with additive solution in single blood bag buckets with filter pack for Thermo Scientific™ HAEMAFlex™6/8 rotors.
$381.50 - $381.50
Products
Catalog Number Mfr. No. Description Price Quantity    
75-003-842
thermo scientific™
75003842
450-550mL Triple, Quad or Quint Blood Bag with Additive Solution Adapter (set of 2) Pack for $381.50
Description & Specifications
Thermo Scientific™ blood banking centrifuges offer the flexibility to select higher capacity rotors or match existing workflows, with a choice of Thermo Scientific™ HAEMAFlex™ rotors and bucket systems.
Address your lab’s regular- to medium-throughput blood processing needs with the HAEMAFlex 6 swinging bucket rotor with 6 x 550mL blood bag capacity or, utilizing the same bucket system, upgrade to 8 x 550mL bags with the HAEMAFlex 8 swinging bucket rotor.
Select an adapter for 450–550mL Triple, Quad or Quint blood bag with additive solution.