Please login to your online account to display your discounted pricing

Fisher Scientific™ Adapters for Fisher Scientific™ accuSpin™ 4-Place Rotor with Rectangular Bucket


Manufacturer: fisher scientific™  75005387

 View more versions of this product

Catalog No. 13-100-607

  • / Each
 

Description & Specifications

Specifications

Container Type 15mL bottle
Color Brown
For Use With (Equipment) accuSpin 1/1R 4-Place Rotor
Size 17 O.D. x 120mm L