Please login to your online account to display your discounted pricing

Fisher Scientific™ Adapters for Fisher Scientific™ accuSpin™ 4-Place Rotor with Rectangular Bucket


Manufacturer: fisher scientific™  75005330

 View more versions of this product

Catalog No. 13-100-609

  • / Each
 

Description & Specifications

Specifications

Size 13 O.D. x 117mm L
Container Type 7mL bottle
For Use With (Equipment) accuSpin 1/1R 4-Place Rotor
Color Grey