missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tocris™ Adaphostin

p210bcr/abl kinase inhibitor

$181.19 - $762.58

Specifications

Product Type Adaphostin
Inhibitors Adaphostin
Molecular Weight 393.48
Purity >98
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
371610R Tocris™
RDS371610
10mg Each for $181.19
371650R Tocris™
RDS371650
50mg Each for $762.58
Specifications

Description

p210bcr/abl tyrosine kinase inhibitor (IC50 = 14μM). Induces apoptosis in T-lymphoblastic human leukemia cell lines (IC50 values range from 16.8 to 216.3nM) in vitro. Displays selectivity for chronic myelogenous leukemia (CML) myeloid progenitors in vitro.
  • Chemical Name: 4-[[(2,5-Dihydroxyphenyl)methyl]amino]benzoic acid tricyclo[3.3.1.13,7]dec-1-yl ester
  • Formula: C24H27NO4
  • CAS Number: 241127-58-2
  • Solubility: Soluble to 100mM in DMSO and to 100mM in ethanol
  • Purity: >98
Description & Specifications

Specifications

Adaphostin
Adaphostin
393.48
>98