missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Adapalene

$188.04 - $231.43

Chemical Identifiers

CAS 106685-40-9
Molecular Formula C28H28O3
Molecular Weight (g/mol) 412.529
MDL Number MFCD03106112
InChI Key LZCDAPDGXCYOEH-UHFFFAOYSA-N
Synonym 6-3-1-adamantyl-4-methoxyphenyl-2-naphthoic acid, 6-3-adamantan-1-yl-4-methoxyphenyl-2-naphthoic acid, adaferin, adapalene, adapaleno, adapaleno inn-spanish, adapalenum, adapalenum inn-latin, differin, differine
PubChem CID 60164
ChEBI CHEBI:31174
IUPAC Name 6-[3-(1-adamantyl)-4-methoxyphenyl]naphthalene-2-carboxylic acid
SMILES COC1=C(C=C(C=C1)C2=CC3=C(C=C2)C=C(C=C3)C(=O)O)C45CC6CC(C4)CC(C6)C5
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ67025MB
View Documents
Alfa Aesar™
J67025MB
25mg
Each for $188.04
 
AAJ67025MF
View Documents
Alfa Aesar™
J67025MF
50mg
Each for $231.43
 
Description

Description

Specifications

Chemical Identifiers

106685-40-9
412.529
LZCDAPDGXCYOEH-UHFFFAOYSA-N
60164
6-[3-(1-adamantyl)-4-methoxyphenyl]naphthalene-2-carboxylic acid
C28H28O3
MFCD03106112
6-3-1-adamantyl-4-methoxyphenyl-2-naphthoic acid, 6-3-adamantan-1-yl-4-methoxyphenyl-2-naphthoic acid, adaferin, adapalene, adapaleno, adapaleno inn-spanish, adapalenum, adapalenum inn-latin, differin, differine
CHEBI:31174
COC1=C(C=C(C=C1)C2=CC3=C(C=C2)C=C(C=C3)C(=O)O)C45CC6CC(C4)CC(C6)C5

Specifications

White
106685-40-9
MFCD03106112
6-3-1-adamantyl-4-methoxyphenyl-2-naphthoic acid, 6-3-adamantan-1-yl-4-methoxyphenyl-2-naphthoic acid, adaferin, adapalene, adapaleno, adapaleno inn-spanish, adapalenum, adapalenum inn-latin, differin, differine
COC1=C(C=C(C=C1)C2=CC3=C(C=C2)C=C(C=C3)C(=O)O)C45CC6CC(C4)CC(C6)C5
412.529
CHEBI:31174
Solid
25mg
C28H28O3
14,150
LZCDAPDGXCYOEH-UHFFFAOYSA-N
6-[3-(1-adamantyl)-4-methoxyphenyl]naphthalene-2-carboxylic acid
60164
412.53
Adapalene
Safety and Handling

Safety and Handling

P201-P202-P281-P308+P313-P501c

H361

RTECSNumber : QJ1987000

TSCA : No

Recommended Storage : Ambient temperatures

SDS