Please login to your online account to display your discounted pricing

Alfa Aesar™ Adapalene


Quantity 50mg

Manufacturer: alfa aesar™  J67025MF

 View more versions of this product

Catalog No. AAJ67025MF

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 412.53
CAS 106685-40-9
Synonym Retinoic acid receptor ligand; CD271
Physical Form Solid
Chemical Name or Material Adapalene
Color White
RTECS Number QJ1987000
Merck Index 14,150
Quantity 50mg
MDL Number MFCD03106112

Specifications

Chemical Name or Material Adapalene
CAS 106685-40-9
Formula Weight 412.53
MDL Number MFCD03106112
Merck Index 14,150
Synonym Retinoic acid receptor ligand; CD271