missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Adapalene

Manufacturer:  Alfa Aesar™ J67025MB

 View more versions of this product

Catalog No. AAJ67025MB



Specifications

Specifications

Adapalene
25mg
C28H28O3
14,150
LZCDAPDGXCYOEH-UHFFFAOYSA-N
6-[3-(1-adamantyl)-4-methoxyphenyl]naphthalene-2-carboxylic acid
60164
412.53
White
106685-40-9
MFCD03106112
6-3-1-adamantyl-4-methoxyphenyl-2-naphthoic acid, 6-3-adamantan-1-yl-4-methoxyphenyl-2-naphthoic acid, adaferin, adapalene, adapaleno, adapaleno inn-spanish, adapalenum, adapalenum inn-latin, differin, differine
COC1=C(C=C(C=C1)C2=CC3=C(C=C2)C=C(C=C3)C(=O)O)C45CC6CC(C4)CC(C6)C5
412.529
CHEBI:31174
Solid

Chemical Identifiers

106685-40-9
412.529
LZCDAPDGXCYOEH-UHFFFAOYSA-N
60164
6-[3-(1-adamantyl)-4-methoxyphenyl]naphthalene-2-carboxylic acid
C28H28O3
MFCD03106112
6-3-1-adamantyl-4-methoxyphenyl-2-naphthoic acid, 6-3-adamantan-1-yl-4-methoxyphenyl-2-naphthoic acid, adaferin, adapalene, adapaleno, adapaleno inn-spanish, adapalenum, adapalenum inn-latin, differin, differine
CHEBI:31174
COC1=C(C=C(C=C1)C2=CC3=C(C=C2)C=C(C=C3)C(=O)O)C45CC6CC(C4)CC(C6)C5
Safety and Handling

Safety and Handling

P201-P202-P281-P308+P313-P501c

H361

RTECSNumber : QJ1987000

TSCA : No

Recommended Storage : Ambient temperatures

Documents