Please login to your online account to display your discounted pricing

Adamantane-1-carbohydrazide, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material Adamantane-1-carbohydrazide
Assay 97%
Assay Percent Range 97%
Molecular Formula C11H18N2O
Formula Weight 194.28
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

RH01025DA

  SDS
fisher scientific company
RH01025DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 194.28
Quantity 1g
Assay 97%
Chemical Name or Material Adamantane-1-carbohydrazide
Packaging Amber Glass Bottle
Assay Percent Range 97%
Molecular Formula C11H18N2O
CAS 17846-15-0