Please login to your online account to display your discounted pricing

Adamantane-1-carbohydrazide, 97%, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  RH01025DA

 View more versions of this product

Catalog No. RH01025DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 194.28
Quantity 1g
Assay 97%
Chemical Name or Material Adamantane-1-carbohydrazide
Packaging Amber Glass Bottle
Assay Percent Range 97%
Molecular Formula C11H18N2O
CAS 17846-15-0

Specifications

Chemical Name or Material Adamantane-1-carbohydrazide
Assay 97%
Assay Percent Range 97%
Molecular Formula C11H18N2O
Formula Weight 194.28