Please login to your online account to display your discounted pricing

ADA, 98+%, for biochemistry, ACROS Organics™


Quantity 25g
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  AC327680250

 View more versions of this product

Catalog No. AC327680250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.0%
Physical Form Powder
Formula Weight 190.16
Quantity 25g
Color White to almost white
EINECS Number 247-530-0
Purity Grade For biochemistry
TSCA TSCA
Merck Index 15, 135
CAS Max % 100.0
CAS Min % 98.0
Water 1% max. (K.F.)
Synonym N-(2-Acetamido)iminodiacetic acid
Assay Percent Range 98+%
Linear Formula H2NCOCH2N(CH2CO2H)2
CAS 26239-55-4
Chemical Name or Material ADA
Molecular Formula C6H10N2O5
MDL Number MFCD00008031
Packaging Plastic bottle
Infrared Spectrum Authentic

Specifications

Chemical Name or Material ADA
CAS 26239-55-4
CAS Min % 98.0
CAS Max % 100.0
Purity Grade For biochemistry
Assay ≥98.0%
Assay Percent Range 98+%
Molecular Formula C6H10N2O5
Linear Formula H2NCOCH2N(CH2CO2H)2
Formula Weight 190.16
MDL Number MFCD00008031
Synonym N-(2-Acetamido)iminodiacetic acid