Please login to your online account to display your discounted pricing

ADA, 98+%, for biochemistry, ACROS Organics™

Chemical Identifiers

Chemical Name or Material ADA
CAS 26239-55-4
CAS Min % 98.0
CAS Max % 100.0
Purity Grade For biochemistry
Assay ≥98.0%
Assay Percent Range 98+%
Molecular Formula C6H10N2O5
Linear Formula H2NCOCH2N(CH2CO2H)2
Formula Weight 190.16
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC327680250

  SDS
thermo fisher scientific
AC327680250
Plastic bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC327681000

  SDS
fisher scientific acros
327681000
Plastic bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.0%
Physical Form Powder
Formula Weight 190.16
Quantity 25g
Color White to almost white
EINECS Number 247-530-0
TSCA TSCA
Purity Grade For biochemistry
Merck Index 15, 135
CAS Min % 98.0
CAS Max % 100.0
Chemical Name or Material ADA
Assay Percent Range 98+%
Linear Formula H2NCOCH2N(CH2CO2H)2
CAS 26239-55-4
Synonym N-(2-Acetamido)iminodiacetic acid
Water 1% max. (K.F.)
Infrared Spectrum Authentic
Packaging Plastic bottle
Molecular Formula C6H10N2O5
MDL Number MFCD00008031

Biological buffer