missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ ADA, 0.2M buffer soln., pH 7.0

$83.27 - $130.46

Chemical Identifiers

CAS 7415-22-7
Molecular Formula C6H9N2NaO5
Molecular Weight (g/mol) 212.137
MDL Number MFCD00065483
InChI Key QCGKKVWCABJQQI-UHFFFAOYSA-M
PubChem CID 23679048
IUPAC Name sodium;2-[(2-amino-2-oxoethyl)-(carboxymethyl)amino]acetate
SMILES C(C(=O)N)N(CC(=O)O)CC(=O)[O-].[Na+]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ61176AE View Documents Alfa Aesar™
J61176AE
100mL Each for $83.27
 
AAJ61176AK View Documents Alfa Aesar™
J61176AK
250mL Each for $130.46
 
Specifications

Chemical Identifiers

7415-22-7
212.137
QCGKKVWCABJQQI-UHFFFAOYSA-M
sodium;2-[(2-amino-2-oxoethyl)-(carboxymethyl)amino]acetate
C6H9N2NaO5
MFCD00065483
23679048
C(C(=O)N)N(CC(=O)O)CC(=O)[O-].[Na+]

Specifications

7415-22-7
MFCD00065483
QCGKKVWCABJQQI-UHFFFAOYSA-M
sodium;2-[(2-amino-2-oxoethyl)-(carboxymethyl)amino]acetate
23679048
7.0
100mL
C6H9N2NaO5
0.2M buffer soln.
C(C(=O)N)N(CC(=O)O)CC(=O)[O-].[Na+]
212.137
Liquid
Colorless
ADA buffer soln.
Safety and Handling

Safety and Handling

Recommended Storage : 2° to 8°C