missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ ADA, 0.2M buffer soln., pH 6.0

$77.57 - $121.51

Chemical Identifiers

CAS 7415-22-7
Molecular Formula C6H9N2NaO5
Molecular Weight (g/mol) 212.137
MDL Number MFCD00065483
InChI Key QCGKKVWCABJQQI-UHFFFAOYSA-M
PubChem CID 23679048
IUPAC Name sodium;2-[(2-amino-2-oxoethyl)-(carboxymethyl)amino]acetate
SMILES C(C(=O)N)N(CC(=O)O)CC(=O)[O-].[Na+]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ60054AE View Documents Alfa Aesar™
J60054AE
100mL Each for $77.57
 
AAJ60054AK View Documents Alfa Aesar™
J60054AK
250mL Each for $121.51
 
Specifications

Chemical Identifiers

7415-22-7
212.137
QCGKKVWCABJQQI-UHFFFAOYSA-M
sodium;2-[(2-amino-2-oxoethyl)-(carboxymethyl)amino]acetate
C6H9N2NaO5
MFCD00065483
23679048
C(C(=O)N)N(CC(=O)O)CC(=O)[O-].[Na+]

Specifications

7415-22-7
MFCD00065483
QCGKKVWCABJQQI-UHFFFAOYSA-M
sodium;2-[(2-amino-2-oxoethyl)-(carboxymethyl)amino]acetate
23679048
6.0
100mL
C6H9N2NaO5
0.2M buffer soln.
C(C(=O)N)N(CC(=O)O)CC(=O)[O-].[Na+]
212.137
Liquid
Colorless
ADA buffer soln.
Safety and Handling

Safety and Handling

Recommended Storage : 2° to 8°C