missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Acycloguanosine, 98%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC445240500
Specifications
Acycloguanosine | |
256°C to 260°C | |
59277-89-3 | |
C8H11N5O3 | |
9-2-hydroxyethoxy methyl guanine, aciclovir, aciclovirum, acycloguanosine, acyclovir, vipral, virorax, wellcome-248u, zovir, zovirax | |
MKUXAQIIEYXACX-UHFFFAOYSA-N | |
2-amino-9-(2-hydroxyethoxymethyl)-3H-purin-6-one | |
2022 | |
225.2 |
Glass bottle | |
50mg | |
97.5% min. (HPLC) | |
15134 | |
Solubility in water: 0.7mg/mL | |
C1=NC2=C(N1COCCO)NC(=NC2=O)N | |
225.2 | |
CHEBI:2453 | |
98% |
Chemical Identifiers
59277-89-3 | |
225.2 | |
9-2-hydroxyethoxy methyl guanine, aciclovir, aciclovirum, acycloguanosine, acyclovir, vipral, virorax, wellcome-248u, zovir, zovirax | |
CHEBI:2453 | |
C1=NC2=C(N1COCCO)NC(=NC2=O)N |
C8H11N5O3 | |
MKUXAQIIEYXACX-UHFFFAOYSA-N | |
2022 | |
2-amino-9-(2-hydroxyethoxymethyl)-3H-purin-6-one |
Safety and Handling
Causes serious eye irritation, May cause respiratory irritation, Causes skin irritation
Serious eye damage/eye irritation (category 2), Specific target organ toxicity after single exposure (category 3), Skin corrosion/irritation (category 2)
EINECSNumber : 261-685-1
RMF - Research Use or Further Mfg