Invitrogen™ ACVR1 (ALK2) Recombinant Human Protein DFS Item
Activin A receptor, type I (ACVR1) belongs to the transforming growth factor-beta (TGF-beta) superfamily of structurally related signaling proteins.
Manufacturer:  Invitrogen™ LSPV4878
 View more versions of this product
Catalog No. PV4878
Description
Description
ACVR1 signals a specific transcriptional response in concert with activin type II receptors
Specifications
Specifications
NP_001096.1
Frozen
69.1kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Catalytic
Kinase Assay
STKR Kinase Family
Yes
Target & Lead Identification & Validation|Kinase Biology|Biochemical Kinase Assays|Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis|Industrial & Applied Science|Pharma & Biopharma
50mM Tris pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Polysorbate 20, 2mM DTT, 50% glycerol
1mg
RUO - research use only
ACVR1 (ALK2)
Baculovirus
Human
GST Tag
Documents